Learn More
Aviva Systems Biology Corp Cct7 Antibody - C-terminal region (ARP51809_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51809_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cct7. It was validated on Western Blot. . Alternate names: AA408524, AL022769, Ccth, CctzApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-596