Learn More
TARGETMOL CHEMICALS INC CCTA-1523 5MG
SDP
Catalog No. 502578995
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CCTA-1523 is an efflux function of ABCG2 inhibitor. CCTA-1523 selectively reverses ABCG2-mediated MDR in cancer cells. purity: 99%
Catalog No. 50-257-8995 Supplier TARGETMOL CHEMICALS INC Supplier No. T95875MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CCTA-1523 is an efflux function of ABCG2 inhibitor. CCTA-1523 selectively reverses ABCG2-mediated MDR in cancer cells. purity: 99%