Learn More
Abcam CCVJ (9-(2-Carbo x y-2-cyanovinyl)julolidine), Fluorescent molecular rotor, 10MG
Catalog No. 502705262
Encompass_Preferred
MW 268.31 Da, Purity >97%. Fluorescent molecular rotor. Sensitive to viscosity. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5262 Supplier Abcam Supplier No. AB14536810MG
MW 268.31 Da, Purity >97%. Fluorescent molecular rotor. Sensitive to viscosity. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement