Learn More
Abcam CCVJ (9-(2-Carbo x y-2-cyanovinyl)julolidine), Fluorescent molecular rotor, 125MG
Catalog No. 502705261
Encompass_Preferred
MW 268.31 Da, Purity >97%. Fluorescent molecular rotor. Sensitive to viscosity. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5261 Supplier Abcam Supplier No. AB145368125MG
MW 268.31 Da, Purity >97%. Fluorescent molecular rotor. Sensitive to viscosity. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement