Learn More
Medchemexpress LLC Ccx-777 | 1226686-36-7 | 98.5% | 593.78 g/mol | C31H43N7O3S | 10MG
SDP
Catalog No. 5000235446
Encompass_Preferred
CCX-777 is a small-molecule partial agonist of β-arrestin-2 recruitment to ACKR3 (atypical chemokine receptor 3), provided as a purified research compound for in vitro and preclinical studies. The compound has formula C31H43N7O3S and a molecular weight of 593.78 g/mol.
Catalog No. 50-002-35446 Supplier Medchemexpress LLC Supplier No. HY12381310MG
CCX-777 is a small-molecule partial agonist of β-arrestin-2 recruitment to ACKR3 (atypical chemokine receptor 3), provided as a purified research compound for in vitro and preclinical studies. The compound has formula C31H43N7O3S and a molecular weight of 593.78 g/mol.