Learn More
Medchemexpress LLC CCZ01048 TFA | 98.5% | 1622.75 | 5 MG
SDP
Catalog No. 5000423889
Encompass_Preferred
CCZ01048 TFA is an α-MSH analogue that exhibits high binding affinity to melanocortin 1 receptor (MC1R) with a Ki of 0.31 nM. It shows rapid internalization into B16F10 melanoma cells and demonstrates high in vivo stability, making it a promising candidate for PET imaging of malignant melanoma.
  • α-MSH analogue
  • High binding affinity to melanocortin 1 receptor (MC1R) with a Ki of 0.31 nM
  • Rapid internalization into B16F10 melanoma cells
  • High in vivo stability
  • Promising candidate for PET imaging of malignant melanoma
Catalog No. 50-004-23889 Supplier Medchemexpress LLC Supplier No. HYP2336A5MG
CCZ01048 TFA is an α-MSH analogue that exhibits high binding affinity to melanocortin 1 receptor (MC1R) with a Ki of 0.31 nM. It shows rapid internalization into B16F10 melanoma cells and demonstrates high in vivo stability, making it a promising candidate for PET imaging of malignant melanoma.
  • α-MSH analogue
  • High binding affinity to melanocortin 1 receptor (MC1R) with a Ki of 0.31 nM
  • Rapid internalization into B16F10 melanoma cells
  • High in vivo stability
  • Promising candidate for PET imaging of malignant melanoma