Learn More
Cayman Chemical CCZ01048 (trifluoroacetate salt), Purity: ≥98%, 5 mg
SDP
Catalog No. 502942649
Encompass_Preferred
A synthetic peptide; has been conjugated to radioligands for use in PET imaging of tumors expressing MC1R and for tissue distribution studies in vivo
Catalog No. 50-294-2649 Supplier Cayman Chemical Supplier No. 414595 MG
A synthetic peptide; has been conjugated to radioligands for use in PET imaging of tumors expressing MC1R and for tissue distribution studies in vivo