Learn More
Cayman Chemical CCZ01048trIfluoROCTAte sa 10mg
SDP
Catalog No. 502944394
Encompass_Preferred
A synthetic peptide; has been conjugated to radioligands for use in PET imaging of tumors expressing MC1R and for tissue distribution studies in vivo
Catalog No. 50-294-4394 Supplier Cayman Chemical Supplier No. 4145910 MG
A synthetic peptide; has been conjugated to radioligands for use in PET imaging of tumors expressing MC1R and for tissue distribution studies in vivo