Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CD 293 AGT™ Medium

Catalog No. 12529001
Change view
Click to view available options
Quantity:
10 kg
10 L
100 L
Format:
Ropak Bucket
Securitainer Capped Vial
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
12-529-001 100 L Ropak Bucket
12-529-003 10 kg Ropak Bucket
12-529-012 10 L Securitainer Capped Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-529-001 Supplier Gibco™ Supplier No. 12529001
Add to Cart
Add to Cart
Request a Quote

CD 293 AGT™ is a protein-free, chemically-defined medium in an easy-to-use granular format.

CD 293 AGT™ is a protein-free, chemically-defined medium in an easy-to-use granular format. CD 293 AGT™ is optimized for the growth of suspension cultures of 293 cells.

AGT = Advanced Granulation Technology

Specifications

Cell Line 293 (HEK)
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Powder (AGT)
Product Type CD 293 Medium
Serum Level Serum-free
Sterility Non-sterile
With Additives Low Glucose, HEPES, Sodium Pyruvate, Sodium Bicarbonate
Without Additives No Glutamine, No Phenol Red
Glucose Concentration 4500 mg/L
Quantity 100 L
Shipping Condition Wet Ice
Endotoxin Level Low
Format Ropak Bucket
Included Antibiotics No antibiotics
Inorganic Salts Calcium, Magnesium, Sodium Phosphate
Purity or Quality Grade Certified
Reconstituted Volume 100 L
Content And Storage Store at 2°C to 8°C in the dark.
Culture Environment CO2
Culture Type Suspension Cell Culture
Show More Show Less
Can you please provide information as to how many grams of powder are present in your CD 293 AGT Medium products?

1 L (Cat. No. 12529020): 17.9 g/L
10 L (Cat. No. 12529012): 178.8 g/10 L
100 L (Cat. No. 12529001): 1788.4 g/100 L
10 kg (Cat. No. 12529003): 10,000 g/10 kg

What are the main benefits of using AGT?

Complete formulations
- Homogeneous distribution of minute components
- Complete complex formulations in a dry format, including serum-free, protein-free, and chemically defined nutrient media
- Pre-adjusted to the appropriate pH and osmolality
- Require only standard supplementation with L-glutamine for L-glutamine dependent systems
- Can be customized for modified catalog and customers' proprietary formulations

Granular format
- Dissolves instantly for faster media preparation times
- Dust-free granules reduce mess and environmental contamination
- Performance equivalent to liquid media
- Workflow improvement
- Helps reduce total cycle costs due to lower number of components, decreasing time involved in raw material planning, procurement, and testing
- Faster and easier media preparation, with less potential for error
- Easy to scale up from research through production, in batch sizes from 2 kg to 6,000 kg
- Available in various packaging options

What is AGT?

Granulated cell culture media is an easy-to-use, granular form of dry media that is produced by applying fluid bed granulation technology to cell culture nutrient media manufacturing. The patented process allows production of complete formulations of a variety of serum-free, protein-free, and chemically defined media in a dry format.

AGT offers the benefits of liquid media without the cost, storage, and transportation issues. It is a complete medium that is pH and osmolality pre-adjusted. The granules dissolve instantly for faster media preparation time than conventional dry powder media. AGT media can therefore help reduce total cycle costs, decreasing time involved in raw material planning, procurement, and testing as well as media preparation. These benefits are all realized while maintaining comparable cell growth to liquid media.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.