Learn More
Biolegend PE anti-human CD33
Supplier:  Biolegend 366608
Encompass_Preferred
PE anti-human CD33 [P67.6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1481124