Learn More
Leica Biosystems CD 8 BOND 4B12 7ML
Supplier:  Leica Biosystems PA0183
Encompass_Preferred
Antigen BackgroundThe CD8 molecule is composed of two chains and has a molecular weight of 32 kD. It is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 2-35% of human peripheral blood lymphocytes.The CD8 antigen is reported to be detected on natural killer cells, 8% of thymocytes, on a subpopulation of 3% of peripheral blood null cells and 15-3% of bone marrow cells.DisclaimerCD8 is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. NC0801189