Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cd-based Core/shell Quantum Dots With Streptavidin Fluorescence Lambda Em 490 Nm 1 Um

Catalog No. 503126073 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.5 mL
2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-6073 2 mL
50-312-5583 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-312-6073 Supplier Sigma Aldrich Supplier No. OCNQSS4902ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Streptavidin has high binding affinity to biotin or biotin-containing molecules such as proteins, peptides, polysaccharides, oligonucleotides. Streptavidin functionalized quantum dots offer an simple and universal method to conjugate those biotinylated molecules to quantum dots for downstream applications such as flow cytometry, Westen blot, microplate assay, fluorescent imaging with microscope for slide-based assay , microarray assay, bead-based assay, fluorescent sensing.The advantages of using quantum dots as fluorophores include high brightness, narrow and symmetric emission, resistant to photobleaching, etc. These unique features make quantum dots ideal optical probes for highly sensitive multiplexing applications.Product of Ocean Nanotech, LLC.

Specifications

Quantity 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.