Learn More
Julabo USA Inc CD-BC6 heating circulator, 1kW heating power, 6L bath, to 150C, 115V/60Hz
SDP
Catalog No. 501374425
Encompass_Preferred
CORIO™ CD-BC6 Heating Circulator combination with 6L stainless steel bath tank for routine laboratory tasks. Units include bright LED display and easy operation.CORIO™ CD immersion circulator provides temperatures from +20 to +150 °C with 1kW heating power. Integrated pump enables temperature incubation for internal or external bath applications. A lever below the display enables simple adjustment between internal and external flow. Pump capacity: 15 L/min; 5.1 PSI. The integrated USB port provide external PC communication for control / monitoring and data logging.The CORIO™ CD has M16x1 male pump connections and comes with sets of 8 and 12 mm hose barbs. For internal and external temperature applications. High-quality stainless-steel bath tanks with bath lid, drain tap and integrated cooling coil. 115V/60Hz
Catalog No. 50-137-4425 Supplier Julabo USA Inc Supplier No. 9012506.02
CORIO™ CD-BC6 Heating Circulator combination with 6L stainless steel bath tank for routine laboratory tasks. Units include bright LED display and easy operation.CORIO™ CD immersion circulator provides temperatures from +20 to +150 °C with 1kW heating power. Integrated pump enables temperature incubation for internal or external bath applications. A lever below the display enables simple adjustment between internal and external flow. Pump capacity: 15 L/min; 5.1 PSI. The integrated USB port provide external PC communication for control / monitoring and data logging.The CORIO™ CD has M16x1 male pump connections and comes with sets of 8 and 12 mm hose barbs. For internal and external temperature applications. High-quality stainless-steel bath tanks with bath lid, drain tap and integrated cooling coil. 115V/60Hz