Learn More
Julabo USA Inc CORIO CD-BT19 open heating circulator bath, 1kw heater, 19L plastic tank, to 100 C, 115V/60Hz.
SDP
Supplier:  Julabo USA Inc 9012319.02
Encompass_Preferred
CORIO™ CD-BT19 Immersion Heating Circulator Open Bath combinations with transparent bath tank for routine laboratory tasks. Units include bright LED display and easy operation.The CORIO™ CD immersion circulator provides temperatures to 100 °C in plastic. Integrated pump enables temperature incubation for internal or external bath applications. A lever below the display enables simple adjustment between internal and external flow. Pump capacity: 15 L/min; 5.1 PSI. The integrated USB port provide external PC communication for control / monitoring and data logging.The CORIO™ CD has M16x1 male pump connections and comes with sets of 8 and 12 mm hose barbs (pump connections M16x1 male). 19L Plexiglass tank, 1kW heater, 115V/60Hz. Lid sold separately (9970291).
Catalog No. 50-137-4405
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov