Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CD CHO AGT™ Medium

Catalog No. 12490017
Change view
Click to view available options
Quantity:
1 L
10 L
100 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-490-017 1 L
12-490-025 10 L
12-490-001 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-490-017 Supplier Gibco™ Supplier No. 12490017
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

CD CHO AGT™ Medium

CD CHO AGT™ (Advanced Granulation Technology) medium is a protein-free, chemically-defined medium optimized for the growth of Chinese hamster ovary (CHO) cells and expression of recombinant proteins in suspension culture. The AGT™ dry media format is designed to provide increased consistency and productivity across all stages of production from development to commercial manufacturing. CD CHO AGT™ medium contains no proteins or peptide components of animal, plant, or synthetic origin, as well as no undefined lysates or hydrolysates.

When to Use CD CHO Medium
  • Your GS CHO cells are recommended to be grown in CD CHO medium
  • Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

    When to Consider Another Gibco™ Medium

    Consider CD FortiCHO™ medium when:
  • The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
  • Maximum batch culture cell densities and protein titers are needed
  • You have time to adapt cells into a new medium in an effort to maximize titers
  • Cell health at harvest is a priority for downstream processing

    Consider CD OptiCHO™ medium when:
  • You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
  • Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
  • Your CHO cell line is finicky, i.e. hard to grow or adapt

    Consider CD DG44 medium when:
  • You use parental DG44 or DXB11 CHO cells (dhfr-)

    Liquid Media Formulations
    Gibco™ AGT™ media are also available in a liquid format that offers the same performance and formulation as the AGT™ media format. Gibco™ liquid media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their AGT™ media counterparts.

    Design Your Own Media Specific to Your Needs
    Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
  • Specifications

    Cell Line CHO
    Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
    Form Powder (AGT)
    Product Type CD CHO Medium
    Serum Level Serum-free
    With Additives Sodium Bicarbonate
    Without Additives No Glutamine, No Phenol Red, No Sodium Pyruvate, No Thymidine, No Hypoxanthine
    Product Line AGT, Gibco
    Quantity 1 L
    Shipping Condition Wet Ice
    Endotoxin Level ≤10 EU/mL, Low
    Format Securitainer Capped Vial
    Included Antibiotics No antibiotics
    Reconstituted Volume 1 L
    Species Hamster
    Content And Storage Store in refrigerator (2–8°C).
    Culture Type Suspension Cell Culture
    Show More Show Less
    Which serum-free medium should I select for my CHO cells?

    Thermo Fisher Scientific offers multiple serum free media (SFM) and chemically defined (CD) media options for CHO cultures. For adherent cultures, CD CHO (10743-011) and CD OptiCHO medium (12681-011) are robust media; both are protein free and animal origin free (AOF). For adherent cultures, CD CHO-A (097-0182DJ) is a widely used protein free and animal origin free medium. There are other formulations available to suit other research needs; contact Technical Support for further information.

    What are the main benefits of using AGT?

    Complete formulations
    - Homogeneous distribution of minute components
    - Complete complex formulations in a dry format, including serum-free, protein-free, and chemically defined nutrient media
    - Pre-adjusted to the appropriate pH and osmolality
    - Require only standard supplementation with L-glutamine for L-glutamine dependent systems
    - Can be customized for modified catalog and customers' proprietary formulations

    Granular format
    - Dissolves instantly for faster media preparation times
    - Dust-free granules reduce mess and environmental contamination
    - Performance equivalent to liquid media
    - Workflow improvement
    - Helps reduce total cycle costs due to lower number of components, decreasing time involved in raw material planning, procurement, and testing
    - Faster and easier media preparation, with less potential for error
    - Easy to scale up from research through production, in batch sizes from 2 kg to 6,000 kg
    - Available in various packaging options

    What is AGT?

    Granulated cell culture media is an easy-to-use, granular form of dry media that is produced by applying fluid bed granulation technology to cell culture nutrient media manufacturing. The patented process allows production of complete formulations of a variety of serum-free, protein-free, and chemically defined media in a dry format.

    AGT offers the benefits of liquid media without the cost, storage, and transportation issues. It is a complete medium that is pH and osmolality pre-adjusted. The granules dissolve instantly for faster media preparation time than conventional dry powder media. AGT media can therefore help reduce total cycle costs, decreasing time involved in raw material planning, procurement, and testing as well as media preparation. These benefits are all realized while maintaining comparable cell growth to liquid media.

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.