Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CD CHO Medium
GREENER_CHOICE

Catalog No. 10743011
Change view
Click to view available options
Quantity:
0.5 L
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-743-011 0.5 L
10-743-029 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-743-011 Supplier Gibco™ Supplier No. 10743011
Add to Cart
Edge
Add to Cart

CD CHO medium is a protein-free, serum-free, chemically-defined medium optimized for the growth of Chinese hamster ovary (CHO) cells and expression of recombinant proteins in suspension culture. With a proven track record of quality for more than 10 years, CD CHO medium contains no proteins or peptide components of animal, plant, or synthetic origin, as well as no undefined lysates or hydrolysates.

When to Use CD CHO Medium

  • Your GS CHO cells are recommended to be grown in CD CHO medium
  • Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

When to Consider Another Gibco™ Medium

Consider CD FortiCHO™ medium when:

  • The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
  • Maximum batch culture cell densities and protein titers are needed
  • You have time to adapt cells into a new medium in an effort to maximize titers
  • Cell health at harvest is a priority for downstream processing

Consider CD OptiCHO™ medium when:

  • You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
  • Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
  • Your CHO cell line is 'finicky,' i.e. hard to grow or adapt

Consider CD DG44 medium when:

  • You use parental DG44 or DXB11 CHO cells (dhfr-)

Dry Media Formulations

Gibco™ liquid media are available in an AGT™ (Advanced Granulation Technology) dry media format that is designed to provide increased consistency and productivity across all stages of production from development to commercial manufacturing. AGT™ media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their liquid media counterparts.

Design Your Own Media Specific to Your Needs

Whether it's a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs.

Specifications

Cell Line CHO
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Concentration 1 X
Form Liquid
Product Type CD CHO Medium
Serum Level Serum-free
With Additives Sodium Pyruvate, Sodium Bicarbonate
Without Additives No Glutamine, No Phenol Red, No Thymidine, No Hypoxanthine
Product Line Gibco
Quantity 0.5 L
Shipping Condition Room Temperature
Endotoxin Level Low
Format Bottle
Included Antibiotics No antibiotics
Species Hamster
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Suspension Cell Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.