Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CD DG44 Medium
GREENER_CHOICE

Catalog No. 12610010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-610-010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-610-010 Supplier Gibco™ Supplier No. 12610010
Add to Cart
Edge
Add to Cart

CD DG44 Medium

CD DG44 Medium is a chemically defined, protein-free medium optimized for the growth of dihydrofolate reductase deficient (DHFR-) Chinese Hamster Ovary (CHO) cells and expression of recombinant proteins in suspension culture. CD DG44 fully chemically defined formulation contains no lysates, hydrolysates, nor proteins or peptide components of animal, plant, or synthetic origin.

When to Use CD DG44 Medium
  • You use parental DG44 or DXB11 CHO cells (dhfr-)

    When to Consider Another Gibco™ Medium

    Consider CD FortiCHO™ medium when:
  • The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
  • Maximum batch culture cell densities and protein titers are needed
  • You have time to adapt cells into a new medium in an effort to maximize titers
  • Cell health at harvest is a priority for downstream processing

    Consider CD OptiCHO™ medium when:
  • You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
  • Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
  • Your CHO cell line is finicky, i.e. hard to grow or adapt

    Consider CD CHO medium when:
  • Your GS CHO cells are recommended to be grown in CD CHO medium
  • Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

    Design Your Own Media Specific to Your Needs
    Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
  • Specifications

    Cell Line CHO DG44
    Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
    Concentration 1 X
    Form Liquid
    Product Type CD DG44 Medium
    Serum Level Serum-free
    With Additives Sodium Pyruvate, Sodium Bicarbonate, Thymidine, Hypoxanthine
    Without Additives No Glutamine, No Phenol Red
    Product Line Gibco
    Quantity 1 L
    Shipping Condition Room Temperature
    Endotoxin Level Low
    Format Bottle(s)
    Included Antibiotics No antibiotics
    Content And Storage Store in refrigerator (2–8°C) and protect from light.
    Culture Type Suspension Cell Culture
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.