Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CD FortiCHO™ Medium
GREENER_CHOICE

Catalog No. A1148301
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1148301 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1148301 Supplier Gibco™ Supplier No. A1148301
  • PRODUCT PROMOTION
    $208.15
    $178.81 / Each of 1

    Save $29.34

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

CD FortiCHO™ Medium

CD FortiCHO™ medium is specifically designed to offer the highest batch and fed-batch culture performance and yield with recombinant CHO cells in a chemically defined environment. The protein-free, animal origin component-free formulation provides consistency and helps reduce the need to screen for adventitious agents. With CD FortiCHO™ medium cell cultures last 2+ days longer than competitive alternatives, which means decreased HCP (Host Cell Protein) concentration for removal in downstream purification.

CD FortiCHO™ medium is fortified to support maximal recombinant CHO cell growth and protein expression in suspension batch culture, offering 120% greater growth and production than select competitive media with simple glucose supplementation. CD FortiCHO™ medium can provide up to 75% greater fed-batch growth and production over glucose-only fed-batch culture for selected cell lines and applications when used with CD EfficientFeed™ C nutrient supplement.

When to Use CD FortiCHO™ Medium
  • The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
  • Maximum batch culture cell densities and protein titers are needed
  • You have time to adapt cells into a new medium in an effort to maximize titers
  • Cell health at harvest is a priority for downstream processing

    When to Consider Another Gibco™ Medium

    Consider CD OptiCHO™ medium when:
  • You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
  • Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
  • Your CHO cell line is finicky, i.e. hard to grow or adapt

    Consider CD CHO medium when:
  • Your GS CHO cells are recommended to be grown in CD CHO medium
  • Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

    Consider CD DG44 when:
  • You use parental DG44 or DXB11 CHO cells (dhfr-)

    Dry Media Formulations
    For early access to CD FortiCHO™ AGT™ medium, please contact Gibco™ Technical Services.

    Design Your Own Media Specific to Your Needs
    Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
  • Specifications

    Cell Line CHO
    Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
    Concentration 1 X
    Form Liquid
    Product Type CD OptiCHO AGT Medium
    With Additives Sodium Pyruvate, Sodium Bicarbonate
    Without Additives No Glutamine, No Phenol Red
    Product Line Gibco
    Quantity 1 L
    Shipping Condition Room Temperature
    Endotoxin Level Low
    Format Bottle
    Included Antibiotics No antibiotics
    Content And Storage Storage:
    Store in the dark at +2 to +8°C. Guaranteed stable for 12 months when properly stored.
    Culture Type Suspension Cell Culture
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.