Learn More
INNOVEX BIOSCIENCES CD MARKERS MULTI-TUMOR CONTROL SLIDES 25
SDP
Catalog No. 50486693
Encompass_Preferred
CD Marker Panel Multi-Tumor control slides, each slide contains non-Hodgkin's lymphoma, mantle cell lymphoma, tonsil tissues positive for 10 CD markers: CD3, CD5, CD10, CD15, CD20, CD23, CD45, CD45RA, 5RO & cyclin d1; 25 slides per package
Catalog No. 50-486-693 Supplier INNOVEX BIOSCIENCES Supplier No. TS90225
CD Marker Panel Multi-Tumor control slides, each slide contains non-Hodgkin's lymphoma, mantle cell lymphoma, tonsil tissues positive for 10 CD markers: CD3, CD5, CD10, CD15, CD20, CD23, CD45, CD45RA, 5RO & cyclin d1; 25 slides per package