Learn More
Grainger CD-R Disc 700 MB 80 min 52x PK100
Catalog No. 502749120
Encompass_Preferred
The Grainger 14F821 is an ABB manual motor starter that provides overload protection for motors up to 10HP at 460VAC. It features adjustable overload relays, an on/off switch with lockout tag capability, and screw terminal connections. This NEMA rated starter has dimensions of 8-3/4"H x 6-3/4"W x 4-3/4"D and weighs approximately 6 lbs. It can be surface or DIN rail mounted and is cULus listed. The 14F821 helps protect motors from damage due to overheating.
Catalog No. 50-274-9120 Supplier Grainger Supplier No. 14F821
The Grainger 14F821 is an ABB manual motor starter that provides overload protection for motors up to 10HP at 460VAC. It features adjustable overload relays, an on/off switch with lockout tag capability, and screw terminal connections. This NEMA rated starter has dimensions of 8-3/4"H x 6-3/4"W x 4-3/4"D and weighs approximately 6 lbs. It can be surface or DIN rail mounted and is cULus listed. The 14F821 helps protect motors from damage due to overheating.