Learn More
Electron Microscopy Sciences GEL-SLIDE CD Series in Antistatic Hinged Box, 2" x 2"
SDP
Catalog No. 501900248
Encompass
Intended for manual applications in which the devices are loaded and unloaded using tweezers or by fingers. Configured as a 2" carrier stored in a protective plastic hinged box. The Gel coating is applied directly to the surface of the plastic tray. Can be easily removed from the box allowing for convenient handling when loading or unloading fixtures are being used. A removable 2" Gel coated glass slide inside a hinged plastic box. For higher temperatures.
Type: Antistatic Hinged
Size: 2" x 2"
Catalog No. 50-190-0248 Supplier Electron Microscopy Sciences Supplier No. 6490022
Intended for manual applications in which the devices are loaded and unloaded using tweezers or by fingers. Configured as a 2" carrier stored in a protective plastic hinged box. The Gel coating is applied directly to the surface of the plastic tray. Can be easily removed from the box allowing for convenient handling when loading or unloading fixtures are being used. A removable 2" Gel coated glass slide inside a hinged plastic box. For higher temperatures.
Type: Antistatic Hinged
Size: 2" x 2"