Learn More
iQ Biosciences CD1 Mouse PBMCs, 5M cells/vial
Supplier:  iQ Biosciences IQBMPB301
Encompass_Preferred
High-quality, high-viability (> 80%) peripheral blood mononuclear cells (PBMCs) from the CD1 mouse, a commonly-used outbred mouse strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2103916