Learn More
Biocare Medical, LLC CD10 Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CM129AK
Encompass_Preferred
Human CD10 antibody, also known as common acute lymphoblastic leukemia (CALLA), has been shown to react with TdT lymphoblastic leukemia, follicular germinal cell lymphoma, Burkitt lymphoma and chronic myelocytic leukemia. CD10 also marks normal early lymphoid progenitor cells, immature B-cells in adult bone marrow and germinal cells in normal tonsil and normal lymphoid tissue. It is also expressed in some non-lymphoid tissues such as fibroblasts, breast myoepithelium and brush border of kidney. CD10 antibody may be used in a panel for mantle cell lymphoma with Cyclin D1 ( ), CD43 ( ), CD5 ( ), IgM ( ), CD23 (-) and CD10 (-).
Catalog No. 50-825-36