Learn More
Biotium CD100 (Semaphorin-4D)(A8), CF488A conjugate, 0.1mg/mL
Supplier:  Biotium BNC880218500
Encompass_Preferred
CD100 (also known as Semaphorin 4D), is a homodimeric protein comprised of 50 kDa subunits. It is expressed on resting T cells and weakly on other lymphocytes, and its expression is increased on B and T cells after stimulation. CD100 is a cell surface receptor that binds to CD72, Plexin B1 and Plexin B2. It promotes B cell aggregation, cell survival, and cell proliferation.Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Catalog No. 50-252-3919