Learn More
ABclonal Technology CD105/Endoglin Rabbit mAb
SDP
Catalog No. 5000011474
Encompass_Preferred
This gene encodes a homodimeric transmembrane protein which is a major glycoprotein of the vascular endothelium This protein is a component of the transforming growth factor beta receptor complex and it binds to the beta1 and beta3 peptides with high affinity Mutations in this gene cause hereditary hemorrhagic telangiectasia also known as Osler-Rendu-Weber syndrome 1 an autosomal dominant multisystemic vascular dysplasia This gene may also be involved in preeclampsia and several types of cancer Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-11474 Supplier ABclonal Technology Supplier No. A25536100UL
This gene encodes a homodimeric transmembrane protein which is a major glycoprotein of the vascular endothelium This protein is a component of the transforming growth factor beta receptor complex and it binds to the beta1 and beta3 peptides with high affinity Mutations in this gene cause hereditary hemorrhagic telangiectasia also known as Osler-Rendu-Weber syndrome 1 an autosomal dominant multisystemic vascular dysplasia This gene may also be involved in preeclampsia and several types of cancer Alternatively spliced transcript variants encoding different isoforms have been found for this gene