Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7412MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: VCAM1/843. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Human placenta or tonsil. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human VCAM1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 110kDa.Chromosome: 1q21.2. Gene Symbol: VCAM1. Gene ID: 7412. Ensemble: ENSG00000162692. Uniprot: NP_001069.
Catalog No. 50-188-5267