Learn More
AdipoGen CD112R (mouse):Fc (human) (rec.) (3 x 10 µg)
SDP
Supplier:  AdipoGen AG40B01703010
Encompass_Preferred
Protein. The extracellular domain of mouse CD112R (aa 28-165) is fused at the C-terminus to the Fc portion of human IgG1. Source: HEK 293 cells. Lyophilized. Contains PBS. Purity: >90% (SDS-PAGE). CD112 functions as an immune checkpoint regulator by modulating T cell signaling, either as a costimulator of T cell function, or as a coinhibitor of T cells proliferation by binding to the receptors CD226 or TIGIT (T cell Ig and immunoreceptor tyrosine-based inhibitory motif (ITIM) domain), respectively. CD112R is a new coinhibitory receptor of the PVR family for human T cells, inhibiting TCR-mediated signals upon binding to CD112/Nectin-2. CD112R is preferentially expressed in T lymphocytes and NK cells. Cancer cells evade immune responses as a result of an immunosuppressive tumor microenvironment and CD112R, by playing a role in the regulation of the immune system, could be a new attractive cancer immunotherapy target.
Catalog No. 50-205-3267