Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3815MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: C117/370 + KIT/982 + KIT/983. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: Gastrointestinal Stromal Tumor (GIST) or testicular germ cell tumor. Melanocytes in the basal layer of the epidermis, mast cells in the dermis of normal skin. Cellular Staining: Cell surface, cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human CD117 proteins. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ~145kDa.Chromosome: 4q12. Gene Symbol: KIT. Gene ID: 3815. Ensemble: ENSG00000157404. Uniprot: NP_000213.
Catalog No. 50-188-4850