Learn More
OriGene CD117 CytoSection, 25 Slides
SDP
Catalog No. NC3243657
Encompass_Preferred
Transient overexpression of KIT, transcript variant 2, in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC3243657 Supplier OriGene Supplier No. TS413772P5
Transient overexpression of KIT, transcript variant 2, in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack