Learn More
CAPRICO BIOTECHNOLOGIES INC Anti-CD117 PE-Cyanine5 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4035075
Encompass_Preferred
The CapricoBio Anti-CD117 PE-Cyanine5 Antibody is a mouse monoclonal antibody that recognizes human CD117. It is conjugated to PE-Cyanine5 and is suitable for flow cytometry, immunofluorescence microscopy, and immunohistochemistry. This antibody specifically binds to CD117, also known as c-Kit or SCF Receptor, which is a 145 kDa transmembrane receptor tyrosine kinase expressed on hematopoietic stem cells, mast cells, melanocytes, and interstitial cells of Cajal. The Anti-CD117 PE-Cyanine5 Antibody can be used for analyzing or isolating CD117-expressing cell populations by flow cytometry or cell sorting.
Catalog No. 50-227-8998