Learn More
Biogenex Laboratories CD117 : T595 : 6mL
SDP
Supplier:  Biogenex Laboratories AM4235M
Encompass_Preferred
c-kit (CD117) is a transmembrane, tyrosine kinase receptor and protooncogene product which is expressed on numerous diverse fetal and adultcells including hematopoietic cells, mast cells, melanocytes, germ cells,and the interstitial cells of Cajal. Its expression in tumors is also diverse.
Catalog No. 50-197-8853