Learn More
ACROBiosystems Recombinant Protein;Human CD117 / c-kit Protein;HEK293;100UG;CD7,CD117,SCFR,c-Kit,KIT
Supplier:  ACROBiosystems CD7H52H4100UG
Encompass_Preferred
Recombinant Protein;Human CD117 / c-kit Protein;100UG;Human CD117, His Tag (CD7-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 516 (Accession # P10721-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.1 kDa. The protein migrates as 66-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD117, His Tag (Cat. No. CD7-H52H4) at 2 μg/mL (100 μl/well) can bind Biotinylated Mouse SCF (Cat. No. SCF-M8228) with a linear range of 0.8-62 ng/mL (QC tested).
Catalog No. 50-165-8969