Learn More
Enzo Life Sciences CD11a (human) monoclonal antibody (MEM-25) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805061C100
Encompass_Preferred
Clone: MEM-25. Isotype: IgG1. Immunogen: Leukocytes from a patient suffering from a LGL-type leukaemia. Species reactivity: Human. Specificity: Recognizes the α subunit (CD11a; αL integrin) of LFA-1. Applications: Flow Cytometry, IP, FUNC. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 15mM (~0.1%) sodium azide.
Catalog No. 50-201-0049