Learn More
Enzo Life Sciences CD11a Recombinant monoclonal antibody (hu1124) (Human IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4090200
Encompass_Preferred
Clone: hu1124 (Efalizumab). Isotype: IgGκ. Species reactivity: Human. Applications: Flow Cytometry, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2936