Learn More
CAPRICO BIOTECHNOLOGIES INC CD11b Antibody, 4007045
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4007045
Encompass_Preferred
The Caprico CD11b Antibody is a mouse monoclonal antibody that recognizes the CD11b protein. It is produced in mouse ascites fluid and is supplied at a concentration of 1 mg/mL in PBS with 0.1% sodium azide. The antibody can be used for applications such as Western blot, immunoprecipitation, and flow cytometry. It has been tested for reactivity in mouse, rat, human, and monkey samples. The immunogen is a recombinant protein corresponding to amino acids 737-1053 of human CD11b. This antibody can be a useful research tool for studying CD11b expression and function.
Catalog No. 50-227-8984