Learn More
CAPRICO BIOTECHNOLOGIES INC CD11b PerCP Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4007035
Encompass_Preferred
The CD11b PerCP antibody is a mouse monoclonal antibody that specifically binds to CD11b, also known as integrin alpha M. It is conjugated to PerCP and is suitable for flow cytometry applications. This antibody recognizes CD11b expressed on monocytes, macrophages, granulocytes, NK cells and subsets of dendritic cells and T cells. It can be used for analyzing leukocyte populations and immunophenotyping. The antibody is available in 100 tests of 5 μL size.
Catalog No. 50-227-8986