Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3687MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: ITGAX/1284. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b. Positive Control: Human dendritic cells. Human lymph nodes and tonsils. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human ITGAX protein fragment (aa 637-827). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 145kDa.Chromosome: 16p11.2. Gene Symbol: ITGAX. Gene ID: 3687. Ensemble: ENSG00000140678. Uniprot: XP_011544154.
Catalog No. 50-188-4841