Learn More
CAPRICO BIOTECHNOLOGIES INC CD11c Antibody, 4079025
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4079025
Encompass_Preferred
The CD11c Antibody is a mouse monoclonal antibody that recognizes CD11c, a type I transmembrane protein highly expressed on dendritic cells. This antibody is unconjugated and has been tested for use in flow cytometry, immunohistochemistry, and immunoprecipitation applications. It is available in multiple sizes ranging from 100 μg to 1 mg. The antibody is produced in vitro using hybridoma technology and is purified by affinity chromatography. According to the manufacturer, this antibody shows high specificity with minimal cross-reactivity. Overall, this appears to be a high-quality CD11c antibody well-suited for research applications involving dendritic cells.
Catalog No. 50-227-8991