Learn More
Sino Biological Recombinant Human IL-2R beta/CD122 Protein, HPLC-verified
Supplier:  Sino Biological 10696HCCH100
Encompass_Preferred
The mature form of human IL2 Rβ (NP_000869.1) extracellular domain (Met 1-Asp 239) with five amino acids (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-211-6282