Learn More
Sino Biological Recombinant Mouse IL-2R beta/CD122 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 50792M02H100
Encompass_Preferred
A DNA sequence encoding the mouse IL2RB (NP_032394.1) (Met1-Glu240) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-5670