Learn More
Biolegend PE/Cyanine7 anti-human CD126 (IL-6Rα)
Supplier:  Biolegend 352810
Encompass_Preferred
PE/Cyanine7 anti-human CD126 (IL-6Rα) [UV4]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1400909