Learn More
CAPRICO BIOTECHNOLOGIES INC CD13 APC Cyanine7 Antibody, Clone WM15, 100 tests
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4038095
Encompass_Preferred
The Caprico CD13 APC Cyanine7 Antibody is a mouse monoclonal antibody that targets human CD13. It is conjugated to APC-Cyanine7 and is supplied as 100 tests at 20 μl per test. The antibody is clone WM15 and has been tested by flow cytometry. CD13 is a 150 kDa cell surface aminopeptidase expressed by myeloid cells, endothelial cells, epithelial cells and fibroblasts. This antibody can be used for flow cytometry analysis of CD13 expression.
Catalog No. 50-227-9011