Learn More
CAPRICO BIOTECHNOLOGIES INC CD13 PE-Cyanine7 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4239085
Encompass_Preferred
The Caprico CD13 PE-Cyanine7 Antibody is a mouse monoclonal antibody that recognizes human CD13, also known as aminopeptidase N. It is conjugated to PE-Cyanine7 and is suitable for flow cytometry, immunofluorescence microscopy and other fluorescent detection methods. This antibody recognizes the 150 kDa CD13 antigen and has been tested for specificity in human, monkey, canine, porcine and rodent samples. It is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. The recommended dilution for flow cytometry is 1:50.
Catalog No. 50-227-9007