Learn More
CAPRICO BIOTECHNOLOGIES INC CD13 PerCP-Cyanine5.5 WM-47 ASR
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4038065
Encompass_Preferred
The Caprico CD13 PerCP-Cyanine5.5 WM-47 ASR is a monoclonal antibody reagent designed to identify human CD13 antigen in flow cytometry applications. It is conjugated to PerCP-Cyanine5.5 and has an excitation max of 482nm and emission max of 678nm. The clone is WM-47. This reagent is intended for laboratory use in qualified procedures to aid in the identification and analysis of cell surface antigens by flow cytometry. It is not intended for therapeutic or diagnostic use. The product should be stored at 2-8°C and protected from prolonged exposure to light. When used properly, it enables identification of CD13 positive cells.
Catalog No. 50-227-9012