Learn More
Sino Biological CD131 / CSF2RB / IL3RB / IL5RB Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10516MM11100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD131 / CSF2RB / IL3RB / IL5RB (rh CD131 / CSF2RB / IL3RB / IL5RB; Catalog#10516-H08H; NP_000386.1; Met 1-Trp 443). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8515