Learn More
Sino Biological Recombinant Human Transferrin Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10516MM01100
Encompass_Preferred
A DNA sequence encoding the human transferrin (NP_001054.1) (Met 1-Pro 698) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-8513