Learn More
ACROBiosystems ACROBiosystems Human CD133 Protein, His Tag
Supplier:  ACROBiosystems CD3H52H420UG
Encompass_Preferred
Recombinant Protein;20UG;Human CD133, His Tag (CD3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - His 865 (Accession # O43490-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 97.2 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD133, His Tag (Cat. No. CD3-H52H4) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CD133 (133-3) Antibody, Human IgG1 with a linear range of 1-16 ng/mL (QC tested).
Catalog No. 50-195-6626