Learn More
Enzo Life Sciences CD134 (human) : Fc (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX522022C050
Encompass_Preferred
Alternative name: OX40, TNFRSF 4. Concentration: 1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The cysteine-rich region of human CD134 (OX40) (aa 29-214) is fused to the Fc portion of human IgG1. Specificity: Binds human and mouse CD134L (OX40L). Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9371