Learn More
Enzo Life Sciences CD134L (soluble) (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX5220233010
Encompass_Preferred
Alternative name: OX40L, TNFSF 4, CD252. Concentration: 0.1mg/ml after reconstitution. Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of human CD134L (OX40L) (aa 52-183) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG-tag. Specificity: Binds to human CD134 (OX40). Long Term Storage: -80°C. Applications: ELISA .
Catalog No. 50-200-9372