Learn More
Sino Biological CD136/MST1R Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11608MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD136/MST1R (rh CD136/MST1R; Catalog#11608-H08H; Q04912; Met1-Leu571). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7739